Learn More
Carnabio Usa Inc BRAF(V600E)/5UG/CATALYTICBRAF
Supplier:  Carnabio Usa Inc 091445
Encompass
BRAF [V600E], Catalytic domain, Mutant [V600E], Amino Acid 433-726, NP_004324.2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0345