Learn More
Medchemexpress LLC Bragsin2 | 342795-08-8 | 99.9% | 289.16 | 10 MM * 1 ML
SDP
Catalog No. 5000346845
Encompass_Preferred
Bragsin2 is a potent, selective, and noncompetitive nucleotide exchange factor BRAG2 inhibitor, with an IC50 of 3 μM. It functions by binding at the interface between the PH domain of BRAG2 and the lipid bilayer, which prevents BRAG2 from activating lipidated Arf GTPase. Bragsin2 has been observed to affect breast cancer stem cells.
  • Potent, selective, and noncompetitive BRAG2 inhibitor.
  • IC50 of 3 μM against BRAG2.
  • Inhibits Arf GTPase activation in HeLa cells.
  • Inhibits the growth of SUM149 and S68 cells.
  • Affects breast cancer stem cells.
Catalog No. 50-003-46845 Supplier Medchemexpress LLC Supplier No. HY111550100MG
Bragsin2 is a potent, selective, and noncompetitive nucleotide exchange factor BRAG2 inhibitor, with an IC50 of 3 μM. It functions by binding at the interface between the PH domain of BRAG2 and the lipid bilayer, which prevents BRAG2 from activating lipidated Arf GTPase. Bragsin2 has been observed to affect breast cancer stem cells.
  • Potent, selective, and noncompetitive BRAG2 inhibitor.
  • IC50 of 3 μM against BRAG2.
  • Inhibits Arf GTPase activation in HeLa cells.
  • Inhibits the growth of SUM149 and S68 cells.
  • Affects breast cancer stem cells.