Learn More
CPC Scientific H-Phe-Asn-Leu-Pro-Leu-Gly-Asn-Tyr-Lys-Lys-Pro-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502109396
Encompass
ONE-LETTER SEQUENCE: FNLPLGNYKKPMOLECULAR FORMULA: C62H95N15O15MOLECULAR WEIGHT:1290.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [62031-54-3], RESEARCH AREA: CancerREFERENCES: G. Gimenez-Gallego et al., Science, 230, 1385 (1985)
Catalog No. 50-210-9396 Supplier CPC Scientific Supplier No. GRFF003A
ONE-LETTER SEQUENCE: FNLPLGNYKKPMOLECULAR FORMULA: C62H95N15O15MOLECULAR WEIGHT:1290.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [62031-54-3], RESEARCH AREA: CancerREFERENCES: G. Gimenez-Gallego et al., Science, 230, 1385 (1985)