Learn More
CPC Scientific H-His-Ala-Glu-Lys-His-Trp-Phe-Val-Gly-Leu-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109401
Encompass
ONE-LETTER SEQUENCE: HAEKHWFVGLMOLECULAR FORMULA: C59H82N16O13MOLECULAR WEIGHT:1223.40STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [198542-00-6]span>
SYNONYMS: FGF acidic I (102-111) (bovine brain)
RESEARCH AREA: Cancer
REFERENCES:
G. Gimenez-Gallego et al., Science, 230, 1385 (1985),
Catalog No. 50-210-9401 Supplier CPC Scientific Supplier No. GRFF005B
ONE-LETTER SEQUENCE: HAEKHWFVGLMOLECULAR FORMULA: C59H82N16O13MOLECULAR WEIGHT:1223.40STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [198542-00-6]span>
SYNONYMS: FGF acidic I (102-111) (bovine brain)
RESEARCH AREA: Cancer
REFERENCES:
G. Gimenez-Gallego et al., Science, 230, 1385 (1985),