Learn More
CPC Scientific H-Pro-Ala-Leu-Pro-Glu-Asp-Gly-Gly-Ser-Gly-Ala-Phe-Pro-Pro-Gly-His-Phe-Lys-Asp-Pro-Lys-Arg-Leu-Tyr-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502109398
Encompass
ONE-LETTER SEQUENCE: PALPEDGGSGAFPPGHFKDPKRLYMOLECULAR FORMULA: C118H173N31O33MOLECULAR WEIGHT:2553.9STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [62031-54-3], RESEARCH AREA: CancerREFERENCES: G. Gimenez-Gallego et al., Science, 230, 1385 (1985)
Catalog No. 50-210-9398 Supplier CPC Scientific Supplier No. GRFF004A
ONE-LETTER SEQUENCE: PALPEDGGSGAFPPGHFKDPKRLYMOLECULAR FORMULA: C118H173N31O33MOLECULAR WEIGHT:2553.9STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [62031-54-3], RESEARCH AREA: CancerREFERENCES: G. Gimenez-Gallego et al., Science, 230, 1385 (1985)