Learn More
Creative Biomart Brain-Derived Neurotrophic Factor (BDNF) Protein
Catalog No. 502084289
Encompass_Preferred
Brain-derived Neurotrophic Factor Human Recombinant produced in E.Coli is a homodimer, non-glycosylated, polypeptide chain containing 2 x 119 amino acids (and an N-terminal Met) and having a total molecular mass of 27kDa.
Catalog No. 50-208-4289 Supplier Creative Biomart Supplier No. THP0238
Brain-derived Neurotrophic Factor Human Recombinant produced in E.Coli is a homodimer, non-glycosylated, polypeptide chain containing 2 x 119 amino acids (and an N-terminal Met) and having a total molecular mass of 27kDa.