Learn More
MOLTOX Molecular Toxicology Inc Brain Heart Infusion Agar, 20 mL/plate, 20 plates/sleeve, Store at Room Temperature
SDP
Catalog No. 50948678
Encompass_Preferred
Also known as BHI, Brain Heart Infusion Agar is a general purpose media used for the cultivation of fastidous and nonfastidous microorganisms. This includes aerobic and anaerobic bacteria. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-678 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S62
Also known as BHI, Brain Heart Infusion Agar is a general purpose media used for the cultivation of fastidous and nonfastidous microorganisms. This includes aerobic and anaerobic bacteria. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.