Learn More
NEOGEN Brain Heart Infusion Agar, 500 g
SDP
Catalog No. 502015387
Encompass_Preferred
Brain Heart Infusion Agar is used for the cultivation of a wide variety of fastidious organisms and is not intended for use in the diagnosis of disease or other conditions in humans.A general purpose nutritious agar base. This medium was first used for the isolation of dental pathogens. With the addition of 7% defibrinated blood the medium will support the growth of a wide range of fastidious organisms, the phosphate buffer will help neutralize the acids produced from the utilization of glucose and thus maintain viability. The medium is not recommended for the determination of hemolytic reactions because of the glucose content.
 For more information visit https://www.neogen.com/solutions/Microbiology/brain-heart-infusion-agar
Catalog No. 50-201-5387 Supplier NEOGEN Supplier No. 700003193
Brain Heart Infusion Agar is used for the cultivation of a wide variety of fastidious organisms and is not intended for use in the diagnosis of disease or other conditions in humans.A general purpose nutritious agar base. This medium was first used for the isolation of dental pathogens. With the addition of 7% defibrinated blood the medium will support the growth of a wide range of fastidious organisms, the phosphate buffer will help neutralize the acids produced from the utilization of glucose and thus maintain viability. The medium is not recommended for the determination of hemolytic reactions because of the glucose content.
 For more information visit https://www.neogen.com/solutions/Microbiology/brain-heart-infusion-agar