Learn More
Andwin Scientific Brain Heart Infusion Agar, 500 g
SDP
Catalog No. NC0733857
Encompass_Preferred
Designed for use with multiple species of bacteria. To prepare media add 52 g of Brain Heart Infusion Agar to a liter of distilled water. Follow instructions. The media will not be sterile until autoclaved prior to use.
Catalog No. NC0733857 Supplier Andwin Scientific Supplier No. B02112
Designed for use with multiple species of bacteria. To prepare media add 52 g of Brain Heart Infusion Agar to a liter of distilled water. Follow instructions. The media will not be sterile until autoclaved prior to use.