Learn More
Bioworld Brain-Heart Infusion Agar, 2 kg
Supplier:  Bioworld 306200123
Encompass_Preferred
Brain-Heart Infusion AgarUsed in the cultivation of a wide variety of microorganisms including bacteria, yeasts and moldsWith the addition of 5% or 10% sheep blood, it is used for the isolation and cultivation of a wide variety of fungal species from clinical and nonclinical sourcespH: 7.4Dehydrated Appearance: beige, homogeneous powderPrepared Appearance: light to medium amber agarSoluble in boiling waterContains:Brain Heart Infusion from Solids Peptic Digest of Animal Tissue Pancreatic Digest of Casein Sodium Chloride Dextrose Disodium Phosphate Agar
Catalog No. 50-254-280