Learn More
Alkali Scientific Brain Heart Infusion Agar (BHI) 20mL, 15x100mm Plates, 10/Pk
SDP
Catalog No. 5000662648
Encompass_Preferred
Brain Heart Infusion (BHI) Agar 20mL Plates, 10/Pk is an enriched, nonselective culture medium formulated to support the growth of a broad range of microorganisms, including fastidious bacteria, yeasts, and molds. Derived from nutrient rich infusions of brain and heart tissue, the medium provides essential growth factors that promote robust microbial recovery. BHI agar is commonly used in clinical diagnostics, research laboratories, and environmental microbiology where broad spectrum cultivation is required. It is particularly effective for organisms such as Streptococcus, Neisseria, and Haemophilus species. Each plate is pre poured with 20mL of sterile agar to maintain consistent depth, reliable growth conditions, and dependable performance during incubation and routine microbiological testing.
Catalog No. 50-006-62648 Supplier Alkali Scientific Supplier No. BHIBT01
Brain Heart Infusion (BHI) Agar 20mL Plates, 10/Pk is an enriched, nonselective culture medium formulated to support the growth of a broad range of microorganisms, including fastidious bacteria, yeasts, and molds. Derived from nutrient rich infusions of brain and heart tissue, the medium provides essential growth factors that promote robust microbial recovery. BHI agar is commonly used in clinical diagnostics, research laboratories, and environmental microbiology where broad spectrum cultivation is required. It is particularly effective for organisms such as Streptococcus, Neisseria, and Haemophilus species. Each plate is pre poured with 20mL of sterile agar to maintain consistent depth, reliable growth conditions, and dependable performance during incubation and routine microbiological testing.