Learn More
Teknova Brain Heart Infusion Agar Plates. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388111
Encompass
Brain Heart Infusion (BHI) broth is a highly nutritive medium and useful for cultivating a variety of microorganisms like aerobic and anaerobic bacteria, yeasts, and molds. BHI broth is commonly used in blood culture work and allied pathological investigations.
Catalog No. 50-238-8111 Supplier Teknova Supplier No. B1010
Brain Heart Infusion (BHI) broth is a highly nutritive medium and useful for cultivating a variety of microorganisms like aerobic and anaerobic bacteria, yeasts, and molds. BHI broth is commonly used in blood culture work and allied pathological investigations.