Learn More
Teknova Brain Heart Infusion Agar Plates. 30ml fill. 100mm. 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387418
Encompass
Brain Heart Infusion (BHI) broth is a highly nutritive medium and useful for cultivating a variety of microorganisms like aerobic and anaerobic bacteria, yeasts, and molds. BHI broth is commonly used in blood culture work and allied pathological investigations.
Catalog No. 50-238-7418 Supplier Teknova Supplier No. 2B13574PK
Brain Heart Infusion (BHI) broth is a highly nutritive medium and useful for cultivating a variety of microorganisms like aerobic and anaerobic bacteria, yeasts, and molds. BHI broth is commonly used in blood culture work and allied pathological investigations.