Learn More
Alkali Scientific Brain Heart Infusion (BHI) Broth, 10/Pk, 0.5mL
SDP
Catalog No. 5000662352
Encompass_Preferred
Brain Heart Infusion (BHI) Broth, 0.5mL Tubes, 10/Pk is a highly nutritious, general purpose growth medium formulated to support cultivation of a wide range of fastidious and non fastidious microorganisms including bacteria, yeasts, and molds. Derived from brain and heart infusions, the enriched formulation promotes reliable microbial recovery in clinical, industrial, and research laboratories. The 0.5mL fill volume is suited for small volume testing, sterility validation, and microbial enrichment workflows. Pre filled sterile tubes simplify handling and reduce preparation time in regulated environments. The nutrient rich composition supports consistent organism growth and recovery, making BHI broth suitable for sterility testing, microbial limit testing, and environmental monitoring where dependable microbial detection and culture performance are essential.
Catalog No. 50-006-62352 Supplier Alkali Scientific Supplier No. BHIBT0005
Brain Heart Infusion (BHI) Broth, 0.5mL Tubes, 10/Pk is a highly nutritious, general purpose growth medium formulated to support cultivation of a wide range of fastidious and non fastidious microorganisms including bacteria, yeasts, and molds. Derived from brain and heart infusions, the enriched formulation promotes reliable microbial recovery in clinical, industrial, and research laboratories. The 0.5mL fill volume is suited for small volume testing, sterility validation, and microbial enrichment workflows. Pre filled sterile tubes simplify handling and reduce preparation time in regulated environments. The nutrient rich composition supports consistent organism growth and recovery, making BHI broth suitable for sterility testing, microbial limit testing, and environmental monitoring where dependable microbial detection and culture performance are essential.