Learn More
Alkali Scientific Brain Heart Infusion (BHI) Broth, 10/Pk, 3mL
SDP
Catalog No. 5000662618
Encompass_Preferred
Brain Heart Infusion (BHI) Broth, 3mL Tubes, 10/Pk is an enriched microbiological growth medium formulated for the cultivation of fastidious and non fastidious microorganisms including bacteria, yeasts, and molds. The formulation, derived from brain and heart infusions, provides a nutrient dense environment that supports reliable microbial recovery in clinical diagnostics, pharmaceutical quality control, and laboratory research. The 3mL fill volume offers flexibility for microbial enrichment, sterility testing, and environmental monitoring applications. Each tube is pre filled and sterilized to simplify preparation and maintain consistency across microbiology workflows. Its broad nutrient composition encourages strong organism growth, making BHI broth a dependable choice for microbial limit testing, culture maintenance, and routine microbiological analysis in regulated laboratory environments.
Catalog No. 50-006-62618 Supplier Alkali Scientific Supplier No. BHIBT003M
Brain Heart Infusion (BHI) Broth, 3mL Tubes, 10/Pk is an enriched microbiological growth medium formulated for the cultivation of fastidious and non fastidious microorganisms including bacteria, yeasts, and molds. The formulation, derived from brain and heart infusions, provides a nutrient dense environment that supports reliable microbial recovery in clinical diagnostics, pharmaceutical quality control, and laboratory research. The 3mL fill volume offers flexibility for microbial enrichment, sterility testing, and environmental monitoring applications. Each tube is pre filled and sterilized to simplify preparation and maintain consistency across microbiology workflows. Its broad nutrient composition encourages strong organism growth, making BHI broth a dependable choice for microbial limit testing, culture maintenance, and routine microbiological analysis in regulated laboratory environments.