Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Brain Heart Infusion Broth with 15% Glycerol
SDP

Catalog No. 23001810 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Dimensions (Dia. x H):
15 x 45 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (Dia. x H)
23-001-810 15 x 45 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-810 Supplier Hardy Diagnostics™ Supplier No. D114
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A cryopreservative for the freezing of microorganisms

  • A variety of supplements have been added to further enhance the recovery of fastidious microorganisms
  • A 15% concentration of glycerol is added to the broth to maintain viability of microorganisms and to protect cell membranes. Glycerol acts as a cryoprotective agent and allows for long-term bacterial preservation

Specifications

Appearance Clear and Amber in Color
Broth Type Brain Heart Infusion
Cap Color Black
Certifications/Compliance ISO 13485 , ISO 17025 , ISO 17025, BSI-ISO 13485
Concentration 15% Glycerol
Additive Glycerol
Format Glass Cryovial
Manufacturing Origin United States
Media Type Broth
Product Type Broth Tube
Formulation Brain Heart Infusion Broth (BHI) with 15% Glycerol
For Use With (Application) Freezing of Microorganisms
Includes Glass Cryovial
Material Glass
No. per Pack 100
Packaging Type Package
Quantity 100 x 2 mL
Volume (Metric) Specimen 2 mL
Dimensions (Dia. x H) 15 x 45 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.