Learn More
Teknova Brain Heart Infusion Broth. 1kg.
SDP
Catalog No. 502387613
Encompass
Brain Heart Infusion (BHI) broth is a highly nutritive medium and useful for cultivating a variety of microorganisms like aerobic and anaerobic bacteria, yeasts, and molds. BHI broth is commonly used in blood culture work and allied pathological investigations.
Catalog No. 50-238-7613 Supplier Teknova Supplier No. B9505
Brain Heart Infusion (BHI) broth is a highly nutritive medium and useful for cultivating a variety of microorganisms like aerobic and anaerobic bacteria, yeasts, and molds. BHI broth is commonly used in blood culture work and allied pathological investigations.