Learn More
Biolog Inc BHI BROTH - Brain Heart Infusion Broth, 10 Tubes/Pkg
SDP
Catalog No. NC0356154
Encompass
This item has a minimum qty of 3 per supplier requirements.
Brain Heart Infusion Broth (BHI BROTH) is an enriched non-selective media intended for the cultivation of most anaerobic bacteria and other fastidious microorganisms. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.BHI BROTH is a general-purpose enriched non-selective broth media that is useful in the cultivation of fastidious and non-fastidious microorganisms from a variety of clinical and non-clinical specimens. It is also used for the preparation of inocula for identification and susceptibility tests, and is especially useful as a blood culture medium. The basic nutritive properties of this media are brain heart infusion from solids and meat peptone. It is supplemented with vitamin K and hemin as growth factors for most anaerobes.
Catalog No. NC0356154 Supplier Biolog Inc Supplier No. AS872
This item has a minimum qty of 3 per supplier requirements.
Brain Heart Infusion Broth (BHI BROTH) is an enriched non-selective media intended for the cultivation of most anaerobic bacteria and other fastidious microorganisms. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.BHI BROTH is a general-purpose enriched non-selective broth media that is useful in the cultivation of fastidious and non-fastidious microorganisms from a variety of clinical and non-clinical specimens. It is also used for the preparation of inocula for identification and susceptibility tests, and is especially useful as a blood culture medium. The basic nutritive properties of this media are brain heart infusion from solids and meat peptone. It is supplemented with vitamin K and hemin as growth factors for most anaerobes.