Learn More
Teknova Brain Heart Infusion Broth. 500mL, Sterile.
SDP
Catalog No. NC9734832
Encompass
Brain Heart Infusion (BHI) broth is a highly nutritive medium and useful for cultivating a variety of microorganisms like aerobic and anaerobic bacteria, yeasts, and molds. BHI broth is commonly used in blood culture work and allied pathological investigations.
This item is not returnable. View return policy
Catalog No. NC9734832 Supplier Teknova Supplier No. B9993
Brain Heart Infusion (BHI) broth is a highly nutritive medium and useful for cultivating a variety of microorganisms like aerobic and anaerobic bacteria, yeasts, and molds. BHI broth is commonly used in blood culture work and allied pathological investigations.