Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Brain Heart Infusion Broth with Ciprofloxacin
SDP

Catalog No. 23001756 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Fill Volume:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
23-001-756 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-756 Supplier Hardy Diagnostics™ Supplier No. K258
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use as a selective pre-enrichment broth for the cultivation of ciprofloxacin-resistant E. coli and other gram negative rods

  • Urologists use this as a rectal screening medium before they perform a prostate gland biopsy
    • The biopsy procedure is known for introducing resistant organisms into the bloodstream from the rectum. This media will allow urologists to screen the men first to see if they will be at risk of a difficult to treat infection or septicemia
  • To be used with plated media.

Specifications

Appearance Clear
Broth Type Brain Heart Infusion
Color Medium Amber
Concentration 10 μg/mL
Additive Ciprofloxacin
Detects Ciprofloxacin-resistant E. coli and other Gram-negative Rods
Product Type Brain Heart Infusion, Broth Tube
Target Organism Class Gram-negative Rods
Test Type Microbial Cultivation
Fill Volume 5 mL
Formulation Brain Heart Infusion Broth with Ciprofloxacin, 10μg/mL
For Use With (Application) Rectal Screening Medium, Cultivation of Ciprofloxacin-resistant E. coli and Other Gram Negative Rods
Material Polycarbonate
No. per Pack 20
Packaging Type Pack
Quantity 20 x 5 mL
Species Escherichia coli
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.