Learn More
Teknova Brain Heart Infusion Broth with Yeast Extract and L-Cysteine. 500mL, Sterile. 8 Pack.
SDP
Catalog No. 502387778
Encompass
Brain Heart Infusion (BHI) broth is a highly nutritive medium and useful for cultivating a variety of microorganisms like aerobic and anaerobic bacteria, yeasts, and molds. BHI broth is commonly used in blood culture work and allied pathological investigations.
Catalog No. 50-238-7778 Supplier Teknova Supplier No. B99708PK
Brain Heart Infusion (BHI) broth is a highly nutritive medium and useful for cultivating a variety of microorganisms like aerobic and anaerobic bacteria, yeasts, and molds. BHI broth is commonly used in blood culture work and allied pathological investigations.