Learn More
Teknova Brain Heart Infusion Modified Broth, 500mL, Sterile. 8 pack.
SDP
Catalog No. 502746009
Encompass
Brain Heart Infusion (BHI) broth is a highly nutritive medium and useful for cultivating a variety of microorganisms like aerobic and anaerobic bacteria, yeasts, and molds. BHI broth is commonly used in blood culture work and allied pathological investigations.
Catalog No. 50-274-6009 Supplier Teknova Supplier No. B99858PK
Brain Heart Infusion (BHI) broth is a highly nutritive medium and useful for cultivating a variety of microorganisms like aerobic and anaerobic bacteria, yeasts, and molds. BHI broth is commonly used in blood culture work and allied pathological investigations.