Learn More
Alzet ALZETTM Brain Infusion Kit 3, Materials for Brain Infusion from any ALZET Osmotic Pump, Low Profile, Wide Base Cannulae (30 ga) Extend 1-3 mm Below Skull Surface
SDP
Catalog No. NC9257658
Encompass
ALZETTM Brain Infusion Kits are designed specifically for use with ALZET pumps for targeted delivery to the central nervous system, such as for infusion into the cerebral ventricles for wider distribution or direct microperfusion of discrete brain structures. ALZET Brain Infusion Kit 3 is optimized to reach the mouse lateral ventricles or other structures of similar depth. The cannulae are a low profile, wider design for stability and extend 1-3 mm from the skull surface.Each ALZET Brain Infusion Kit 3 includes materials for 10 brain infusions:•10 Brain Infusion Cannulae (low profile, wide pedestal, 30 gauge, extend 1-3 mm from skull surface)•10 Vinyl Catheter Tubes•40 Depth-Adjustment Spacers•1 Instruction SheetKit is sterilized by gamma irradiation and ready for use.
Catalog No. NC9257658 Supplier Alzet Supplier No. 0008851
ALZETTM Brain Infusion Kits are designed specifically for use with ALZET pumps for targeted delivery to the central nervous system, such as for infusion into the cerebral ventricles for wider distribution or direct microperfusion of discrete brain structures. ALZET Brain Infusion Kit 3 is optimized to reach the mouse lateral ventricles or other structures of similar depth. The cannulae are a low profile, wider design for stability and extend 1-3 mm from the skull surface.Each ALZET Brain Infusion Kit 3 includes materials for 10 brain infusions:•10 Brain Infusion Cannulae (low profile, wide pedestal, 30 gauge, extend 1-3 mm from skull surface)•10 Vinyl Catheter Tubes•40 Depth-Adjustment Spacers•1 Instruction SheetKit is sterilized by gamma irradiation and ready for use.