Learn More
Electron Microscopy Sciences Brain Matrix Single Razor Blade, 25/pk
SDP
Supplier:  Electron Microscopy Sciences 7093370
Encompass_Preferred
These razor blades are specifically made to work and fit all of our Brain Matrices. They measure 118 mm x 19 mm x .2286 mm. They are packed 25 blades in a box.
Catalog No. 50-930-552