Learn More
Cayman Chemical BraIn NatrIuretIc PeptID32 5mg
SDP
Supplier:  Cayman Chemical 397245 MG
Encompass_Preferred
A natriuretic peptide; binds to natriuretic peptide receptors on rat vascular smooth muscle cells (IC50 = 7.3 nM) and induces cGMP accumulation in the same cells (EC50 = 170 nM); induces relaxation of norepinephrine-precontracted isolated rat aortic strips (EC50 = 6.7 nM); increases urine and urinary electrolyte excretion and induces hypotension in rats at 3 UG/kg
Catalog No. 50-291-9264