Learn More
Medchemexpress LLC Brain Natriuretic Peptide (1-32), rat acetate | 98.7% | 3512.99 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP1519B5MG
Encompass_Preferred
Brain Natriuretic Peptide (1-32), rat acetate (BNP (1-32), rat acetate) is a 32 amino acid polypeptide secreted by the ventricles of the heart in response to excessive stretching of heart muscle cells (cardiomyocytes). This product is intended for research purposes only and not for patient use.
  • Combats cardiac stress
  • Reduces blood pressure and ventricular fibrosis
  • Regulates salt and water homeostasis
  • Stimulates renal and depressor responses
Catalog No. 50-004-23060