Learn More
Genscript Corporation BNP, Human
Supplier:  Genscript Corporation Z027461
Encompass
Natriuretic Peptide Precursor B acts as a cardiac hormone with a variety of biological actions including natriuresis, diuresis, vasorelaxation, and inhibition of renin and aldosterone secretion. It is thought to play a key role in cardiovascular homeostasis. Helps restore the body's salt and water balance. Improves heart function.
Catalog No. 50-276-489