Learn More
Avanti Polar Lipids L-α-phosphatidylinositol-4,5-bisphosphate (Brain, Porcine) (ammonium salt), 383907-42-4, MFCD28901390, 10 mg
Catalog No. NC0998838
Encompass
Formula Weight: 1096.385, Purity >99%, Stability: 3 Months, Storage Temperature: -20°C. Although PI(4,5)P2 is a minor component of cell membranes, it plays a critical role as a substrate for a number of important signaling proteins. PI(4,5)P2 is an intermediate in the IP3/DAG pathway where it is hydrolyzed by phospholipase C to liberate the second messengers, inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG). PI(4,5)P2 is also a substrate for PI 3-kinase where it is phosphorylated to PI(3,4,5)P3, an activator of downstream signaling components such as the protein kinase AKT.
Catalog No. NC0998838 Supplier Avanti Polar Lipids Supplier No. 840046X10MG
Formula Weight: 1096.385, Purity >99%, Stability: 3 Months, Storage Temperature: -20°C. Although PI(4,5)P2 is a minor component of cell membranes, it plays a critical role as a substrate for a number of important signaling proteins. PI(4,5)P2 is an intermediate in the IP3/DAG pathway where it is hydrolyzed by phospholipase C to liberate the second messengers, inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG). PI(4,5)P2 is also a substrate for PI 3-kinase where it is phosphorylated to PI(3,4,5)P3, an activator of downstream signaling components such as the protein kinase AKT.