Learn More
Avanti Polar Lipids BRAIN PL (4 5) P2 IN CHLOR 1MG
Catalog No. NC9926280
Encompass
Brain PI(4,5)P2,L-a-Phosphatidylinositol-4,5-bisphosphate (Brain,Porcine-Triammonium Salt),1 mg, Molecular Weight: 1,098.19,Molecular Formula: C47H94N3O19P3,Percent Composition: C 51.40% H 8.63% N 3.83% O 27.68% P 8.46%
Catalog No. NC9926280 Supplier Avanti Polar Lipids Supplier No. 840046X1MG
Brain PI(4,5)P2,L-a-Phosphatidylinositol-4,5-bisphosphate (Brain,Porcine-Triammonium Salt),1 mg, Molecular Weight: 1,098.19,Molecular Formula: C47H94N3O19P3,Percent Composition: C 51.40% H 8.63% N 3.83% O 27.68% P 8.46%