Learn More
Electron Microscopy Sciences Brain Punch Set of Five, 0.25 to 1.25 mm
SDP
Catalog No. 501926846
Encompass_Preferred
The Brain Punch Kit is a set of electropolished and sharpened punches for the Palkovit's punch technique. Complete with spring loaded nylon expellers and a convenient handle. The nuclei are punched from frozen brain sections on glass slides, and expelled into the reagents. The spring-loaded expellers avoid risk of contamination that might be caused by expelling with breath.
Size: 0.25 to 1.25 mm
Catalog No. 50-192-6846 Supplier Electron Microscopy Sciences Supplier No. 57401
The Brain Punch Kit is a set of electropolished and sharpened punches for the Palkovit's punch technique. Complete with spring loaded nylon expellers and a convenient handle. The nuclei are punched from frozen brain sections on glass slides, and expelled into the reagents. The spring-loaded expellers avoid risk of contamination that might be caused by expelling with breath.
Size: 0.25 to 1.25 mm