Learn More
Electron Microscopy Sciences Brain Punch Individual, 1.75 mm
SDP
Catalog No. 501926849
Encompass_Preferred
Electropolished and sharpened punches for the Palkovit's punch technique.
Size: 1.75 mm
Catalog No. 50-192-6849 Supplier Electron Microscopy Sciences Supplier No. 57404
Electropolished and sharpened punches for the Palkovit's punch technique.
Size: 1.75 mm