Learn More
Enzo Life Sciences Brain (rat), (tissue extract) (200 µg)
SDP
Catalog No. NC1620762
Encompass_Preferred
Formulation: Liquid. In SDS-PAGE sample buffer, pH 6.8, containing 40mM TRIS-HCl, 1% SDS, 50mM DTT, 7.5% glycerol, and 0.003% bromophenol blue. Applications: WB. Shipping: Shipped on Dry Ice. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles.
Catalog No. NC1620762 Supplier Enzo Life Sciences Supplier No. ADILYTRB100F
Formulation: Liquid. In SDS-PAGE sample buffer, pH 6.8, containing 40mM TRIS-HCl, 1% SDS, 50mM DTT, 7.5% glycerol, and 0.003% bromophenol blue. Applications: WB. Shipping: Shipped on Dry Ice. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles.