Learn More
Fine Science Tools Brain Sectioning Knife, Straight, 45 cm, each
SDP
Catalog No. NC1370654
Encompass
Product: Brain Sectioning Knife/45cm/ Blade Shape: Straight/ Cutting Edge: 30cm/ Thickness: 1.7mm/ Alloy / Material: Stainless Steel
Catalog No. NC1370654 Supplier Fine Science Tools Supplier No. 1015230
Product: Brain Sectioning Knife/45cm/ Blade Shape: Straight/ Cutting Edge: 30cm/ Thickness: 1.7mm/ Alloy / Material: Stainless Steel