Learn More
Avanti Polar Lipids Brain Total Lipid Extract (Porcine), 86088-88-2, 100mg
Catalog No. NC1302651
Encompass
Total Lipid Extract is a chloroform:methanol extract of the respective tissue (Brain, Soybean, Heart, etc.). This extract is partitioned against deionized water, and the chloroform phase is concentrated.
Catalog No. NC1302651 Supplier Avanti Polar Lipids Supplier No. 131101C100MG
Total Lipid Extract is a chloroform:methanol extract of the respective tissue (Brain, Soybean, Heart, etc.). This extract is partitioned against deionized water, and the chloroform phase is concentrated.