Learn More
Medchemexpress LLC HY-19620 100mg , Branaplam CAS:1562338-42-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19620100MG
Encompass_Preferred
HY-19620 100mg Branaplam CAS: 1562338-42-4 Branaplam (LMI070; NVS-SM1) is a highly potent, selective and orally active survival motor neuron-2 (SMN2) splicing modulator with an EC50 of 20 nM for SMN. Branaplam inhibits human-ether-a-go-go-related gene (hERG) with an IC50 of 6.3 μM. Branaplam elevates full-length SMN protein and extends survival in a severe spinal muscular atrophy (SMA) mouse model. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9270