Learn More
Medchemexpress LLC Branaplam | 1562338-42-4 | 98.90% | 393.48 | 200 MG
SDP
Catalog No. 5000400794
Encompass_Preferred
Branaplam is a highly potent, selective and orally active survival motor neuron-2 (SMN2) splicing modulator with an EC50 of 20 nM for SMN. It inhibits human-ether-a-go-go-related gene (hERG) with an IC50 of 6.3 μM. Branaplam elevates full-length SMN protein and extends survival in a severe spinal muscular atrophy (SMA) mouse model.
  • Highly potent, selective and orally active survival motor neuron-2 (SMN2) splicing modulator.
  • Elevates full-length SMN protein.
  • Extends survival in a severe spinal muscular atrophy (SMA) mouse model.
  • EC50 of 20 nM for SMN.
  • IC50 of 6.3 μM for hERG.
Catalog No. 50-004-00794 Supplier Medchemexpress LLC Supplier No. HY19620200MG
Branaplam is a highly potent, selective and orally active survival motor neuron-2 (SMN2) splicing modulator with an EC50 of 20 nM for SMN. It inhibits human-ether-a-go-go-related gene (hERG) with an IC50 of 6.3 μM. Branaplam elevates full-length SMN protein and extends survival in a severe spinal muscular atrophy (SMA) mouse model.
  • Highly potent, selective and orally active survival motor neuron-2 (SMN2) splicing modulator.
  • Elevates full-length SMN protein.
  • Extends survival in a severe spinal muscular atrophy (SMA) mouse model.
  • EC50 of 20 nM for SMN.
  • IC50 of 6.3 μM for hERG.