Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, HDPE, Tee, Hose Barb to Threaded Adapter, 1"" ID x 1"" ID x 3/4"" NPT(M); 100/PK
Catalog No. 50630499
Encompass
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
Catalog No. 50-630-499 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3062784
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.