Learn More
Kyfora Bio Branched PEI MW 2000 100g
Catalog No. 5000285300
Encompass_Preferred
Branched Polyethylenimine Mw 2000 (bPEI 2000) is branched liquid water soluble polyamine with a high cationic charge density that readily binds anionic substrates It can be used in the formulation of nanocarriers such as PEI pcoated graphene oxide nanoparticles or PEI coated mesoporous silica nanocarriers for a combination of nucleic acid and drug delivery
Catalog No. 50-002-85300 Supplier Kyfora Bio Supplier No. 06089100
Branched Polyethylenimine Mw 2000 (bPEI 2000) is branched liquid water soluble polyamine with a high cationic charge density that readily binds anionic substrates It can be used in the formulation of nanocarriers such as PEI pcoated graphene oxide nanoparticles or PEI coated mesoporous silica nanocarriers for a combination of nucleic acid and drug delivery