Learn More
MSC Brand 3M PTFE Glass Cloth Tape, 5.3 mil, 2 in
Catalog No. 5000803398
Encompass
This general purpose tape is made from a light brown woven glass cloth backing impregnated with PTFE, designed to create a slick, non-stick surface on process machinery. It features a pressure-sensitive silicone adhesive, enabling it to perform effectively at elevated temperatures and function as an anti-stick or easy-release medium.
  • Glass cloth reinforced backing provides high tensile strength and resistance to tears and punctures.
  • Silicone adhesive offers high-temperature performance and long aging.
  • Creates a non-stick, low friction surface on heat sealing and packaging equipment.
  • Performs across a wide temperature range, from -100°F to 500°F.
  • Very low moisture absorption suitable for humid environments.
  • Excellent chemical resistance for challenging applications.
Catalog No. 50-008-03398 Supplier MSC Supplier No. 20721650
This general purpose tape is made from a light brown woven glass cloth backing impregnated with PTFE, designed to create a slick, non-stick surface on process machinery. It features a pressure-sensitive silicone adhesive, enabling it to perform effectively at elevated temperatures and function as an anti-stick or easy-release medium.
  • Glass cloth reinforced backing provides high tensile strength and resistance to tears and punctures.
  • Silicone adhesive offers high-temperature performance and long aging.
  • Creates a non-stick, low friction surface on heat sealing and packaging equipment.
  • Performs across a wide temperature range, from -100°F to 500°F.
  • Very low moisture absorption suitable for humid environments.
  • Excellent chemical resistance for challenging applications.