Learn More
BRANDTECH™ BRAND accu-jet pro Pipette Controller
Catalog No. 13689203 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Color:
Dark Blue
Green
Magenta
Royal Blue
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
13-689-203 Royal Blue
13-689-200 Dark Blue
13-689-201 Magenta
13-689-202 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-689-203 Supplier BRANDTECH™ Supplier No. 26303
The BRAND accu-jet pro allows fast, comfortable filling of serological and volumetric pipettes for cell culture and other applications.
Touch sensitive buttons allow intuitive speed control for fine meniscus control. Additional control for small volume pipettes is provided by a speed limiter thumbwheel.
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips 0.1-200mL volumetric and serological pipets
Color Royal Blue
Model Royal Blue
Type Pipette Controller
Includes acuu-jet pro pipette controller, nickel metal hydride battery, two battery compartment covers, wall support, AC adapter, and two spare 0.2um sterile membrane filters
Length (Metric) 23 cm
Width (Metric) 23 cm
Height (Metric) 6.2 cm
Electrical Requirements CEE plug
Voltage 100/240 V
Frequency 50/60 Hz