Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Empty TipBoxes with Carrier Plates
GREENER_CHOICE

Catalog No. 01670716 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01-670-716 Blue
01-670-713 Gray
01-670-714 Yellow
01-670-715 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 01-670-716 Supplier BRANDTECH™ BRAND™ Supplier No. 732996
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Refillable polypropylene boxes designed to hold BRAND™pipette tips. Carrier plates are color-coded for easy recognition.

  • Polypropylene boxes are highly translucent for easy identification of contents
  • Hinged lid can be rotated 180 degrees to act as lift-off lid

Box Options

  • Gray carrier plate designed to hold nano-cap™ or ultra-micro/crystal tips
  • Yellow carrier plate designed to hold tips fitting 200 μL pipettes
  • Green carrier plate to hold tips fitting 300 μL pipettes
  • Taller box with a blue carrier plate to hold 1000 μL tips
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Color Blue
Holds Micropipette Tips (1000 μL)
Material Polypropylene
Positions 96
Shape Rectangular
Stackable Yes
Sterility Non-sterile
For Use With (Equipment) BRAND 1000 μL pipette tips
Length (Metric) 130 mm
Includes Box, lid, and carrier plate
Product Type Empty Box Reload
Array 8 x 12
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.