Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRANDplates hydroGrade™ 96-Well Microplates

Catalog No. 13882228 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Color:
Clear
Well Shape:
Flat Bottom (F-well)
Round Bottom (U-well)
Total Volume (Metric) Well:
330 μL
350 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Well Shape Total Volume (Metric) Well
13-882-228 Clear Flat Bottom (F-well) 350 μL
13-882-227 Clear Round Bottom (U-well) 330 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-882-228 Supplier BRANDTECH™ Supplier No. 781782
Add to Cart
Add to Cart

Featuring a strongly hydrophilic surface for binding of hydrophilic molecules or for homogeneous assays with hydrophobic molecules that remain in solution

  • Immunoassay surface with strong affinity to hydrophilic molecules, such as glycoproteins and peptides, antibodies with predominantly hydrophilic regions, and nucleic acids
  • Blue alphanumeric embossing for easy well identification
  • Non-sterile

Order Info

Packaging is 20 stacks of 5 plates each (100 plates). Each stack comes with one lid.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Hydrophilic
Certifications/Compliance ANSI, SLAS
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Product Type 96-well Microplate
Material Polystyrene
Volume (Metric) Well 350 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment hydroGrade
Total Volume (Metric) Well 350 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) Immobilization of hydrophilic molecules
Quantity 100/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.