Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRANDplates lipoGrade™ 96-Well Microplates

Catalog No. 13882236 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Color:
Clear
Well Shape:
Flat Bottom (F-well)
Round Bottom (U-well)
Round, Conical Bottom (V-well)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Well Shape
13-882-236 Clear Flat Bottom (F-well)
13-882-234 Clear Round Bottom (U-well)
13-882-237 Clear Round, Conical Bottom (V-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-882-236 Supplier BRANDTECH™ Supplier No. 781842
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Immunoassay plate optimized for the immobilization of hydrophobic molecules

  • High binding for molecules with predominately hydrophobic regions such as lipoproteins or peptides
  • PS, non-sterile
  • Blue alphanumeric embossing on clear, white and black solid plates for easy well identification
  • For solid phase assays with hydrophobic molecules and liquid phase with hydrophilic molecules, respectively
  • Alternative to immunoGrade™ surface
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance ANSI, SLAS
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type High Binding
Color Clear
Product Type 96-well Microtitration Plate
Material Polystyrene
Volume (Metric) Well 350 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment lipoGrade
Well Shape Flat Bottom (F-well)
For Use With (Application) Solid phase with hydrophobic molecules and liquid phase with hydrophilic molecules
Quantity 100/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.