Learn More
Sigma Aldrich Fine Chemicals Biosciences BRAND(R) pipette withdraw volume 2 mL (including bulb), graduations, 0.5 mL, drop quantity 22-26 , per mL | 500EA
Catalog No. NC1756414
Encompass
BRAND(R) pipette withdraw volume 2 mL (including bulb), graduations, 0.5 mL, drop quantity 22-26 , per mL | 500EA
Catalog No. NC1756414 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BR747765500EA
BRAND(R) pipette withdraw volume 2 mL (including bulb), graduations, 0.5 mL, drop quantity 22-26 , per mL | 500EA