Learn More
Sigma Aldrich Fine Chemicals Biosciences BRAND Pipette, 3.5 mL, pack of 500
Catalog No. NC1756415
Encompass
Disposable. Very good reproducibility of the number of drops per mL. Ideal for aliquots. Can be deep-frozen in filled state or changed into a closed vessel by heat-sealing the tip. The integrated pipetting bulb depresses easily, minimize fatigue from frequent pipetting.
Catalog No. NC1756415 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3317671PAK
Disposable. Very good reproducibility of the number of drops per mL. Ideal for aliquots. Can be deep-frozen in filled state or changed into a closed vessel by heat-sealing the tip. The integrated pipetting bulb depresses easily, minimize fatigue from frequent pipetting.