Learn More
Sigma Aldrich Fine Chemicals Biosciences BRAND pipette, 3 mL, 500 pack
Catalog No. NC1756412
Encompass
Disposable. Very good reproducibility of the number of drops per mL. Ideal for aliquots. Can be deep-frozen in filled state or changed into a closed vessel by heat-sealing the tip. The integrated pipetting bulb depresses easily, minimize fatigue from frequent pipetting.
Catalog No. NC1756412 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3317401PAK
Disposable. Very good reproducibility of the number of drops per mL. Ideal for aliquots. Can be deep-frozen in filled state or changed into a closed vessel by heat-sealing the tip. The integrated pipetting bulb depresses easily, minimize fatigue from frequent pipetting.