Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Pipette Tips, 0.5 to 20 μL

Catalog No. 13889153TU Shop All BrandTech Scientific Products
Change view
Click to view available options
Volume (Metric):
0.5 to 20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13-889-153TU 0.5 to 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-889-153TU Supplier BRANDTECH™ BRAND™ Supplier No. 732104TU
Add to Cart
Add to Cart

Engineered to fit the Transferpette™ line of micropipettes

  • Ultra low retention tips
  • Available either in bulk packages or in convenient rack-and-stack formats
  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface thus produced has low surface tension
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain detergents such as Triton™ X-100, SDS, Tween, etc.
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging material properties
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.5 to 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Autoclavable
Clean Claims DNase, RNase, ATP, and Endotoxin Free
Format Refill
Tip Style Narrow
Packaging TipRack
Product Type Pipette Tips
Disposable Limited-use
For Use With (Equipment) Universal Fit Ultramicro
Graduated Yes
Material Polypropylene
Graduations 2 μL, 10 μL
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.