Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ 384-Well Skirted Microplates, BIO-CERT™ PCR QUALITY

Catalog No. 1388276 Shop All BrandTech Scientific Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
13-882-76 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-882-76 Supplier BRANDTECH™ BRAND™ Supplier No. 781345
Only null left
Add to Cart
Add to Cart

Optimized for reliable and precise results in a wide range of PCR applications.

BRAND™ PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:

  • Exceptional Quality and Purity
    • BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
    • Free of contaminants that could interfere with reagents and samples
    • Ultra-smooth vessel interiors minimize enzyme nucleic acid binding
  • Precision Thermal Performance
    • Thin, uniform walls ensure consistent, precise thermal transfer for reliable results
    • Designed for rapid and accurate heat transfer, reducing cycle times
  • Wide Compatibility
    • Suitable for most commercially available thermal cyclers
    • White PCR plates, enhanced with titanium dioxide (TiO2), optimize fluorescence signal reflection and are recommended for qPCR applications
  • User-Friendly Features
    • Blue alphanumeric coding for easy sample identification
    • Well rims designed to prevent cross-contamination and ensure reliable sealing with tailored mats
    • Skirted plates provide extra rigidity, ideal for automated pipetting systems and transport
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Untreated
Format Standard
Array 16 x 24
Autoclavable Autoclavable
Packaging Bagged
Color Clear
Disposable Single-use
Product Type PCR 384-well plate
For Use With (Equipment) Thermal Cyclers
Skirt Style Fully skirted
Material Polypropylene
Volume (Metric) Well 0.03 mL
No. of Wells 384
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 0.03 mL
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) PCR
No. per Pack 50
Quantity 50/Pk.
RNase Free Yes
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.