Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ 1000 μL Blue Pipet Tips

Catalog No. 13688643 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
1000 μL Blue Pipet Tips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
13-688-643 1000 μL Blue Pipet Tips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-688-643 Supplier BRANDTECH™ BRAND™ Supplier No. 712210

missing translation for 'validMainframeMsg'

Precision-molded from high purity, DiHEMDA- and oleamide-free, virgin polypropylene

Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.

  • Uses cadmium-free pigments, all other tips are pigment-free
  • Available either in bulk packages or in convenient rack-and-stack formats
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 to 1000 μL
Sterility Non-sterile
Color Blue
Clean Claims DiHEMDA- and oleamide-free
For Use With (Application) For clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility
Format Rack
Packaging 1 polycarbonate tip box with sliding lid; 60 tips
Product Line Brand
Material Polypropylene
Graduations 250, 500, 1000μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.