Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ BRANDplates cellGrade™ premium, 96-Well Flat-Bottom Microplates, PS

Catalog No. 13882257 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
13-882-257 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-882-257 Supplier BRANDTECH™ BRAND™ Supplier No. 782082
Add to Cart
Add to Cart

cellGrade™ premium surface treatment is ideal for cultivating demanding cell lines in serum-free or serum-reduced environments. Provides a reliable alternative to biologically coated surfaces with good room temperature shelf life.

  • Poly-D-Lysine-equivalent surface delivers similar cell growth and morphology
  • Optimized cell adhesion reduces damage from frequent washing
  • Suitable for for serum-free and serum-reduced cultivation
  • Physical/chemical treatment offers a longer shelf life at room temperature compared to biological coatings
  • Sterilized by B-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Complies with Ph. Eur. and USP 29 standards
  • BIO-CERT™ CELL CULTURE QUALITY: DNA-, DNase-, RNase-free and non-cytoxic
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification

Order Info

Packs of 50 individually wrapped plates with lids

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance ISO 11137, USP 29
Lid With Lid
Array 8 x 12
Packaging 1/Pk., 50/Cs.
Color Clear
Product Type 96-well plate
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment cellGrade Premium
Total Volume (Metric) Well 350 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) Cultivation of cell lines with the highest demands on their environment
No. per Case 50/Cs.
No. per Pack 1/Pk.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.